Assay Design



Gene Transcripts

Gene Symbol
ZCRB1
Entrez Gene ID
85437
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Gene Aliases
MADP-1, MADP1, RBM36, SNRNP31, ZCCHC19
Chromosome Location
Chr.12: 42312086 - 42326130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033114.3 NP_149105.3 2 - 3 293 142
GenBank mRNA AB062361.1 - 2 - 3 293 142
AK308244.1 - 2 - 3 163 142
BC005099.1 - 2 - 3 243 142
BC010177.2 - 2 - 3 258 142
BC022543.1 - 2 - 3 268 142

View Full Product Details