Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282719.1 | NP_001269648.1 | 3 - 4 | 436 | 99 |
NM_001282720.1 | NP_001269649.1 | 3 - 4 | 418 | 99 | |
NM_001282721.1 | NP_001269650.1 | 3 - 4 | 406 | 99 | |
NM_001282722.1 | NP_001269651.1 | 2 - 3 | 262 | 99 | |
NM_021925.3 | NP_068744.1 | 4 - 5 | 562 | 99 | |
XM_011533014.2 | XP_011531316.1 | 4 - 5 | 589 | 99 | |
GenBank mRNA | AK025473.1 | - | 4 - 5 | 500 | 99 |
AK299555.1 | - | 3 - 4 | 407 | 99 | |
AK316162.1 | - | 3 - 4 | 365 | 99 | |
AK316204.1 | - | 2 - 3 | 233 | 99 | |
AK316310.1 | - | 3 - 4 | 389 | 99 | |
BC017473.1 | - | 4 - 5 | 519 | 99 |