Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005260641.3 | XP_005260698.1 | 5 - 6 | 1261 | 99 |
XM_005260643.3 | XP_005260700.1 | 4 - 5 | 928 | 99 | |
XM_005260644.3 | XP_005260701.1 | 3 - 4 | 611 | 99 | |
XM_017027581.1 | XP_016883070.1 | 6 - 7 | 1569 | 99 | |
XM_017027585.1 | XP_016883074.1 | 4 - 5 | 1361 | 99 | |
GenBank mRNA | BC025286.1 | - | 3 - 4 | 601 | 99 |
BC075835.1 | - | 3 - 4 | 601 | 99 | |
BC131747.1 | - | 3 - 4 | 585 | 99 | |
Y10376.1 | - | 5 - 6 | 1240 | 99 |