Assay Design
Gene Transcripts
- Gene Symbol
- DNAJC11
- Entrez Gene ID
-
55735
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C11
- Gene Aliases
-
dJ126A5.1
- Chromosome Location
-
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC014145.2 |
- |
10 - 11 |
1105 |
70 |
View Full Product Details