Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002616.2 | NP_002607.2 | 11 - 12 | 1624 | 80 |
XM_005256689.1 | XP_005256746.1 | 11 - 12 | 1624 | 80 | |
XM_005256690.1 | XP_005256747.1 | 11 - 12 | 1624 | 80 | |
GenBank mRNA | AB002107.1 | - | 10 - 11 | 1386 | 80 |
AB088477.1 | - | 11 - 12 | 1566 | 80 | |
AF022991.1 | - | 11 - 12 | 1573 | 80 | |
AK291061.1 | - | 11 - 12 | 1584 | 80 | |
AK295410.1 | - | 11 - 12 | 1344 | 80 | |
BC028207.1 | - | 10 - 11 | 1539 | 80 | |
BC137346.1 | - | 10 - 11 | 1533 | 80 | |
BC144593.1 | - | 10 - 11 | 1533 | 80 |