Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002616.2 | NP_002607.2 | 19 - 20 | 3307 | 70 |
XM_005256689.1 | XP_005256746.1 | 19 - 20 | 3307 | 70 | |
XM_005256690.1 | XP_005256747.1 | 18 - 19 | 3127 | 70 | |
GenBank mRNA | AB002107.1 | - | 18 - 19 | 3069 | 70 |
AB088477.1 | - | 19 - 20 | 3249 | 70 | |
AF022991.1 | - | 19 - 20 | 3256 | 70 | |
AK291061.1 | - | 19 - 20 | 3267 | 70 | |
BC137346.1 | - | 18 - 19 | 3216 | 70 |