Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002616.2 | NP_002607.2 | 3 - 4 | 609 | 61 |
XM_005256689.1 | XP_005256746.1 | 3 - 4 | 609 | 61 | |
XM_005256690.1 | XP_005256747.1 | 3 - 4 | 609 | 61 | |
GenBank mRNA | AB002107.1 | - | 2 - 3 | 371 | 61 |
AB088477.1 | - | 3 - 4 | 551 | 61 | |
AF022991.1 | - | 3 - 4 | 558 | 61 | |
AK291061.1 | - | 3 - 4 | 569 | 61 | |
AK295410.1 | - | 3 - 4 | 329 | 61 | |
BC028207.1 | - | 2 - 3 | 524 | 61 | |
BC072458.1 | - | 2 - 3 | 522 | 61 | |
BC137346.1 | - | 2 - 3 | 518 | 61 | |
BC144593.1 | - | 2 - 3 | 518 | 61 |