Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164883.1 | NP_001158355.1 | 11 - 12 | 1817 | 67 |
NM_001282597.2 | NP_001269526.1 | 11 - 12 | 1817 | 67 | |
NM_001282598.1 | NP_001269527.1 | 11 - 12 | 1670 | 67 | |
NM_001282599.1 | NP_001269528.1 | 5 - 6 | 758 | 67 | |
NM_001282600.1 | NP_001269529.1 | 5 - 6 | 701 | 67 | |
NM_001320810.1 | NP_001307739.1 | 5 - 6 | 701 | 67 | |
NM_004389.3 | NP_004380.2 | 11 - 12 | 1817 | 67 | |
XM_011532555.2 | XP_011530857.1 | 11 - 12 | 1664 | 67 | |
XM_011532556.2 | XP_011530858.1 | 12 - 13 | 1860 | 67 | |
XM_011532557.2 | XP_011530859.1 | 3 - 4 | 406 | 67 | |
XM_017003403.1 | XP_016858892.1 | 12 - 13 | 1860 | 67 | |
XM_017003404.1 | XP_016858893.1 | 11 - 12 | 1755 | 67 | |
XM_017003405.1 | XP_016858894.1 | 11 - 12 | 1755 | 67 | |
XM_017003406.1 | XP_016858895.1 | 5 - 6 | 791 | 67 | |
GenBank mRNA | AK127226.1 | - | 15 - 16 | 2263 | 67 |
AK295181.1 | - | 11 - 12 | 1616 | 67 | |
AK295493.1 | - | 5 - 6 | 688 | 67 | |
AK303035.1 | - | 11 - 12 | 1670 | 67 | |
BC040458.1 | - | 10 - 11 | 1489 | 67 | |
BC052996.1 | - | 11 - 12 | 1751 | 67 | |
BX537769.1 | - | 5 - 6 | 758 | 67 | |
HQ589336.1 | - | 5 - 6 | 689 | 67 | |
M94151.2 | - | 10 - 11 | 1568 | 67 |