Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257542.2 | XP_005257599.1 | 1 - 2 | 154 | 86 |
XM_005257543.2 | XP_005257600.1 | 1 - 2 | 154 | 86 | |
XM_011525065.1 | XP_011523367.1 | 1 - 2 | 165 | 86 | |
XM_011525068.1 | XP_011523370.1 | 1 - 2 | 72 | 86 | |
XM_017024900.1 | XP_016880389.1 | 1 - 2 | 154 | 86 | |
GenBank mRNA | AB040886.1 | - | 1 - 2 | 47 | 86 |
AK001671.1 | - | 1 - 2 | 165 | 86 | |
AK022913.1 | - | 1 - 2 | 72 | 86 | |
AK225155.1 | - | 1 - 2 | 72 | 86 | |
BC016487.1 | - | 1 - 2 | 131 | 86 | |
BC027992.1 | - | 1 - 2 | 68 | 86 | |
BC038983.1 | - | 1 - 2 | 139 | 86 |