Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321291.1 | NP_001308220.1 | 6 - 7 | 988 | 60 |
XM_005257542.2 | XP_005257599.1 | 6 - 7 | 1012 | 60 | |
XM_005257543.2 | XP_005257600.1 | 6 - 7 | 1012 | 60 | |
XM_011525065.1 | XP_011523367.1 | 6 - 7 | 1023 | 60 | |
XM_011525066.2 | XP_011523368.1 | 6 - 7 | 885 | 60 | |
XM_011525067.2 | XP_011523369.1 | 6 - 7 | 885 | 60 | |
XM_011525068.1 | XP_011523370.1 | 6 - 7 | 926 | 60 | |
XM_011525069.2 | XP_011523371.1 | 6 - 7 | 885 | 60 | |
XM_011525071.2 | XP_011523373.1 | 6 - 7 | 885 | 60 | |
XM_017024900.1 | XP_016880389.1 | 6 - 7 | 1012 | 60 | |
XM_017024901.1 | XP_016880390.1 | 4 - 5 | 547 | 60 | |
GenBank mRNA | AB040886.1 | - | 6 - 7 | 905 | 60 |
AK001671.1 | - | 6 - 7 | 1023 | 60 | |
AK022913.1 | - | 6 - 7 | 930 | 60 | |
AK225426.1 | - | 6 - 7 | 887 | 60 | |
AY169386.1 | - | 4 - 5 | 687 | 60 | |
BC016487.1 | - | 6 - 7 | 989 | 60 | |
BC027992.1 | - | 6 - 7 | 926 | 60 | |
BC071582.1 | - | 6 - 7 | 988 | 60 |