Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000352.4 | NP_000343.2 | 28 - 29 | 3622 | 77 |
NM_001287174.1 | NP_001274103.1 | 28 - 29 | 3625 | 77 | |
XM_011520331.2 | XP_011518633.1 | 28 - 29 | 3647 | 77 | |
XM_017018197.1 | XP_016873686.1 | 28 - 29 | 3685 | 77 | |
XM_017018198.1 | XP_016873687.1 | 28 - 29 | 3682 | 77 | |
XM_017018199.1 | XP_016873688.1 | 28 - 29 | 3690 | 77 | |
XM_017018200.1 | XP_016873689.1 | 28 - 29 | 3640 | 77 | |
XM_017018201.1 | XP_016873690.1 | 28 - 29 | 3685 | 77 | |
XM_017018202.1 | XP_016873691.1 | 27 - 28 | 3486 | 77 | |
XM_017018204.1 | XP_016873693.1 | 22 - 23 | 2598 | 77 | |
GenBank mRNA | AB209084.1 | - | 27 - 28 | 3466 | 77 |
AF087138.1 | - | 28 - 29 | 3588 | 77 | |
L78207.1 | - | 28 - 29 | 3591 | 77 |