Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000352.4 | NP_000343.2 | 5 - 6 | 897 | 62 |
NM_001287174.1 | NP_001274103.1 | 5 - 6 | 897 | 62 | |
XM_017018197.1 | XP_016873686.1 | 5 - 6 | 891 | 62 | |
XM_017018198.1 | XP_016873687.1 | 5 - 6 | 891 | 62 | |
XM_017018201.1 | XP_016873690.1 | 5 - 6 | 891 | 62 | |
XM_017018202.1 | XP_016873691.1 | 4 - 5 | 678 | 62 | |
XM_017018203.1 | XP_016873692.1 | 5 - 6 | 891 | 62 | |
GenBank mRNA | AB209084.1 | - | 5 - 6 | 871 | 62 |
AF087138.1 | - | 5 - 6 | 863 | 62 | |
L78207.1 | - | 5 - 6 | 866 | 62 | |
U63421.1 | - | 5 - 6 | 828 | 62 |