Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 11 - 12 | 1490 | 66 |
NM_015386.2 | NP_056201.2 | 11 - 12 | 1490 | 66 | |
XM_011522981.2 | XP_011521283.1 | 12 - 13 | 1650 | 66 | |
GenBank mRNA | AB088369.1 | - | 12 - 13 | 1626 | 66 |
AK022874.1 | - | 11 - 12 | 1470 | 66 | |
AK026435.1 | - | 7 - 8 | 66 | ||
AK096557.1 | - | 9 - 10 | 1208 | 66 | |
AK293235.1 | - | 8 - 9 | 1034 | 66 | |
AK297557.1 | - | 12 - 13 | 1433 | 66 | |
AL050101.1 | - | 7 - 8 | 792 | 66 | |
BC006306.2 | - | 8 - 9 | 1006 | 66 | |
BC013347.2 | - | 7 - 8 | 798 | 66 | |
BC027726.2 | - | 11 - 12 | 1464 | 66 | |
BC072438.1 | - | 11 - 12 | 1470 | 66 | |
BX648010.1 | - | 9 - 10 | 2158 | 66 |