Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 5 - 6 | 753 | 91 |
NM_015386.2 | NP_056201.2 | 5 - 6 | 753 | 91 | |
XM_011522981.2 | XP_011521283.1 | 6 - 7 | 913 | 91 | |
GenBank mRNA | AB088369.1 | - | 5 - 6 | 730 | 91 |
AK022874.1 | - | 5 - 6 | 733 | 91 | |
AK096557.1 | - | 3 - 4 | 471 | 91 | |
AK297557.1 | - | 6 - 7 | 696 | 91 | |
AL050101.1 | - | 1 - 2 | 55 | 91 | |
AU125729.1 | - | 2 - 3 | 231 | 91 | |
BC006306.2 | - | 2 - 3 | 269 | 91 | |
BC013347.2 | - | 1 - 2 | 61 | 91 | |
BC027726.2 | - | 5 - 6 | 727 | 91 | |
BC072438.1 | - | 5 - 6 | 733 | 91 | |
BC128144.1 | - | 5 - 6 | 730 | 91 | |
BC128145.1 | - | 5 - 6 | 730 | 91 | |
BX648010.1 | - | 4 - 5 | 655 | 91 |