Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005385.3 | NP_005376.2 | 7 - 8 | 554 | 122 |
XM_005265173.1 | XP_005265230.1 | 7 - 8 | 549 | 122 | |
XM_006713171.1 | XP_006713234.1 | 7 - 8 | 549 | 122 | |
XM_006713173.2 | XP_006713236.1 | 8 - 9 | 571 | 122 | |
XM_017006474.1 | XP_016861963.1 | 7 - 8 | 549 | 122 | |
XM_017006477.1 | XP_016861966.1 | 2 - 3 | 1731 | 122 | |
XM_017006478.1 | XP_016861967.1 | 2 - 3 | 1731 | 122 | |
XM_017006479.1 | XP_016861968.1 | 2 - 3 | 1731 | 122 | |
GenBank mRNA | AK126234.1 | - | 2 - 3 | 1731 | 122 |
BC144518.1 | - | 6 - 7 | 2247 | 122 | |
L04288.2 | - | 7 - 8 | 595 | 122 |