Assay Design



Gene Transcripts

Gene Symbol
NKTR
Entrez Gene ID
4820
Gene Name
natural killer cell triggering receptor
Gene Aliases
p104
Chromosome Location
Chr.3: 42600655 - 42648741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005385.3 NP_005376.2 8 - 9 687 88
XM_005265173.1 XP_005265230.1 8 - 9 682 88
XM_006713171.1 XP_006713234.1 8 - 9 682 88
XM_006713173.2 XP_006713236.1 9 - 10 704 88
XM_011533747.2 XP_011532049.1 9 - 10 4016 88
XM_011533750.2 XP_011532052.1 10 - 11 698 88
XM_017006474.1 XP_016861963.1 8 - 9 682 88
XM_017006475.1 XP_016861964.1 9 - 10 686 88
XM_017006476.1 XP_016861965.1 3 - 4 5210 88
XM_017006477.1 XP_016861966.1 3 - 4 1864 88
XM_017006478.1 XP_016861967.1 3 - 4 1864 88
XM_017006479.1 XP_016861968.1 3 - 4 1864 88
XM_017006480.1 XP_016861969.1 10 - 11 698 88
GenBank mRNA AB209888.1 - 7 - 8 5935 88
AK126234.1 - 3 - 4 1864 88
AK292017.1 - 10 - 11 716 88
BC072680.1 - 10 - 11 686 88
BC144518.1 - 7 - 8 2380 88
BX640751.1 - 2 - 3 3227 88
BX641127.1 - 1 - 2 4122 88
BX957215.1 - 1 - 2 1107 88
BX957218.1 - 10 - 11 713 88
CR749415.1 - 1 - 2 1107 88
CR749571.1 - 10 - 11 713 88
L04288.2 - 8 - 9 728 88

View Full Product Details