Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022841.5 | NP_073752.5 | 9 - 9 | 7060 | 113 |
XM_005254603.3 | XP_005254660.2 | 10 - 10 | 7154 | 113 | |
XM_011521925.2 | XP_011520227.1 | 9 - 9 | 6992 | 113 | |
XM_017022506.1 | XP_016877995.1 | 10 - 10 | 9877 | 113 | |
XM_017022507.1 | XP_016877996.1 | 9 - 9 | 7120 | 113 | |
XM_017022508.1 | XP_016877997.1 | 8 - 8 | 8289 | 113 | |
GenBank mRNA | AL832584.1 | - | 1 - 1 | 5870 | 113 |
AL833367.1 | - | 1 - 1 | 5287 | 113 | |
BC112936.1 | - | 7 - 7 | 6880 | 113 |