Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007038.4 | NP_008969.2 | 1 - 2 | 1833 | 68 |
XM_011529448.2 | XP_011527750.1 | 1 - 2 | 1833 | 68 | |
GenBank mRNA | AF142099.1 | - | 1 - 2 | 1226 | 68 |
AK094057.1 | - | 1 - 2 | 1832 | 68 | |
BC093775.1 | - | 1 - 2 | 1188 | 68 | |
BC093777.1 | - | 1 - 2 | 1188 | 68 |