Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005340.1 | NP_001005340.1 | 1 - 2 | 228 | 69 |
NM_002510.2 | NP_002501.1 | 1 - 2 | 228 | 69 | |
XM_005249578.2 | XP_005249635.1 | 1 - 2 | 719 | 69 | |
XM_017011676.1 | XP_016867165.1 | 1 - 2 | 719 | 69 | |
XM_017011677.1 | XP_016867166.1 | 1 - 2 | 719 | 69 | |
XM_017011678.1 | XP_016867167.1 | 1 - 2 | 719 | 69 | |
GenBank mRNA | AF322909.1 | - | 1 - 2 | 126 | 69 |
AJ505015.1 | - | 1 - 2 | 140 | 69 | |
AK292078.1 | - | 1 - 2 | 150 | 69 | |
AK296779.1 | - | 1 - 2 | 150 | 69 | |
AK297051.1 | - | 1 - 2 | 159 | 69 | |
AK297055.1 | - | 1 - 2 | 157 | 69 | |
AK303071.1 | - | 1 - 2 | 156 | 69 | |
AK304671.1 | - | 2 - 3 | 209 | 69 | |
AK312279.1 | - | 1 - 2 | 157 | 69 | |
AY359124.1 | - | 1 - 2 | 148 | 69 | |
BC011595.1 | - | 1 - 2 | 116 | 69 | |
BC025297.1 | - | 1 - 2 | 118 | 69 | |
BC032783.1 | - | 1 - 2 | 144 | 69 | |
BM750843.1 | - | 1 - 2 | 238 | 69 | |
BT007074.1 | - | 1 - 2 | 67 | 69 | |
CD105306.1 | - | 1 - 2 | 216 | 69 | |
X76534.1 | - | 1 - 2 | 158 | 69 |