Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278208.1 | NP_001265137.1 | 9 - 10 | 821 | 92 |
NM_007068.3 | NP_008999.2 | 12 - 13 | 1020 | 92 | |
XM_006724112.2 | XP_006724175.1 | 10 - 11 | 945 | 92 | |
XM_011529834.1 | XP_011528136.1 | 12 - 13 | 1110 | 92 | |
XM_011529835.2 | XP_011528137.1 | 12 - 13 | 991 | 92 | |
XM_011529837.1 | XP_011528139.1 | 10 - 11 | 945 | 92 | |
XM_011529838.1 | XP_011528140.1 | 9 - 10 | 903 | 92 | |
XM_017028542.1 | XP_016884031.1 | 9 - 10 | 903 | 92 | |
GenBank mRNA | AK292617.1 | - | 12 - 13 | 1020 | 92 |
AK297664.1 | - | 9 - 10 | 821 | 92 | |
BC125163.1 | - | 12 - 13 | 917 | 92 | |
BC125164.1 | - | 12 - 13 | 918 | 92 | |
CR456486.1 | - | 12 - 13 | 888 | 92 | |
D63882.1 | - | 12 - 13 | 909 | 92 | |
D64108.1 | - | 12 - 13 | 886 | 92 |