Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278208.1 | NP_001265137.1 | 10 - 11 | 937 | 65 |
NM_007068.3 | NP_008999.2 | 13 - 14 | 1136 | 65 | |
XM_006724112.2 | XP_006724175.1 | 11 - 12 | 1061 | 65 | |
XM_017028542.1 | XP_016884031.1 | 10 - 11 | 1019 | 65 | |
GenBank mRNA | AK292617.1 | - | 13 - 14 | 1136 | 65 |
AK297664.1 | - | 10 - 11 | 937 | 65 | |
BC125163.1 | - | 13 - 14 | 1033 | 65 | |
BC125164.1 | - | 13 - 14 | 1034 | 65 | |
CR456486.1 | - | 13 - 14 | 1004 | 65 | |
D63882.1 | - | 13 - 14 | 1025 | 65 | |
D64108.1 | - | 13 - 14 | 1002 | 65 |