Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007068.3 | NP_008999.2 | 1 - 2 | 152 | 147 |
XM_006724112.2 | XP_006724175.1 | 1 - 2 | 242 | 147 | |
XM_011529834.1 | XP_011528136.1 | 1 - 2 | 242 | 147 | |
XM_011529837.1 | XP_011528139.1 | 1 - 2 | 242 | 147 | |
XM_011529838.1 | XP_011528140.1 | 1 - 2 | 242 | 147 | |
XM_017028542.1 | XP_016884031.1 | 1 - 2 | 242 | 147 | |
GenBank mRNA | AK292617.1 | - | 1 - 2 | 152 | 147 |
BC035658.1 | - | 1 - 2 | 140 | 147 | |
BM545092.1 | - | 1 - 2 | 165 | 147 |