Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019852.4 | NP_062826.2 | 5 - 6 | 1267 | 60 |
XM_006720206.3 | XP_006720269.1 | 5 - 6 | 1168 | 60 | |
XM_011536968.1 | XP_011535270.1 | 5 - 6 | 1262 | 60 | |
GenBank mRNA | AK000222.1 | - | 5 - 6 | 1190 | 60 |
AK093509.1 | - | 3 - 4 | 1049 | 60 | |
BC001650.2 | - | 5 - 6 | 1201 | 60 | |
BC003031.1 | - | 5 - 6 | 1201 | 60 | |
BC052244.1 | - | 5 - 6 | 1151 | 60 | |
BX247964.1 | - | 5 - 6 | 708 | 60 |