Assay Design



Gene Transcripts

Gene Symbol
METTL3
Entrez Gene ID
56339
Gene Name
methyltransferase like 3
Gene Aliases
IME4, M6A, MT-A70, Spo8
Chromosome Location
Chr.14: 21498131 - 21511375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019852.4 NP_062826.2 6 - 7 1458 135
XM_006720206.3 XP_006720269.1 6 - 7 1359 135
XM_011536968.1 XP_011535270.1 6 - 7 1453 135
GenBank mRNA AK000222.1 - 6 - 7 1381 135
AK093509.1 - 4 - 5 1240 135
BC001650.2 - 6 - 7 1392 135
BC003031.1 - 6 - 7 1392 135
BC052244.1 - 6 - 7 1342 135
BX247964.1 - 6 - 7 899 135

View Full Product Details