Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 13 - 14 | 1317 | 120 |
NM_001317963.1 | NP_001304892.1 | 13 - 14 | 1489 | 120 | |
NM_007062.2 | NP_008993.1 | 13 - 14 | 1461 | 120 | |
GenBank mRNA | AK223162.1 | - | 13 - 14 | 1385 | 120 |
AK290681.1 | - | 13 - 14 | 1302 | 120 | |
AK296252.1 | - | 13 - 14 | 1318 | 120 | |
AK297961.1 | - | 12 - 13 | 1146 | 120 | |
BC001652.2 | - | 13 - 14 | 1380 | 120 | |
L07758.1 | - | 13 - 14 | 1380 | 120 |