Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317963.1 | NP_001304892.1 | 1 - 2 | 237 | 67 |
NM_007062.2 | NP_008993.1 | 1 - 2 | 237 | 67 | |
GenBank mRNA | AK223162.1 | - | 1 - 2 | 161 | 67 |
AK290681.1 | - | 1 - 2 | 78 | 67 | |
AK297961.1 | - | 1 - 2 | 189 | 67 | |
BC000067.2 | - | 1 - 2 | 120 | 67 | |
BC001652.2 | - | 1 - 2 | 156 | 67 | |
BC010921.1 | - | 1 - 2 | 171 | 67 | |
BC032127.2 | - | 1 - 2 | 159 | 67 | |
BC040135.1 | - | 1 - 2 | 135 | 67 | |
BC046144.1 | - | 1 - 2 | 151 | 67 | |
DC380488.1 | - | 1 - 2 | 237 | 67 | |
L07758.1 | - | 1 - 2 | 156 | 67 |