Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 2 - 3 | 157 | 83 |
NM_001317963.1 | NP_001304892.1 | 2 - 3 | 301 | 83 | |
NM_007062.2 | NP_008993.1 | 2 - 3 | 301 | 83 | |
GenBank mRNA | AK223162.1 | - | 2 - 3 | 225 | 83 |
AK290681.1 | - | 2 - 3 | 142 | 83 | |
AK296252.1 | - | 2 - 3 | 158 | 83 | |
BC000067.2 | - | 2 - 3 | 184 | 83 | |
BC001652.2 | - | 2 - 3 | 220 | 83 | |
BC010921.1 | - | 2 - 3 | 235 | 83 | |
BC032127.2 | - | 2 - 3 | 223 | 83 | |
BC040135.1 | - | 2 - 3 | 199 | 83 | |
BC046144.1 | - | 2 - 3 | 215 | 83 | |
DC380488.1 | - | 2 - 3 | 301 | 83 | |
L07758.1 | - | 2 - 3 | 220 | 83 |