Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 8 - 9 | 822 | 68 |
NM_001317963.1 | NP_001304892.1 | 8 - 9 | 994 | 68 | |
NM_007062.2 | NP_008993.1 | 8 - 9 | 966 | 68 | |
GenBank mRNA | AK223162.1 | - | 8 - 9 | 890 | 68 |
AK290681.1 | - | 8 - 9 | 807 | 68 | |
AK296252.1 | - | 8 - 9 | 823 | 68 | |
AK297961.1 | - | 7 - 8 | 651 | 68 | |
BC001652.2 | - | 8 - 9 | 885 | 68 | |
L07758.1 | - | 8 - 9 | 885 | 68 |