Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014868.4 | NP_055683.3 | 2 - 3 | 840 | 81 |
XM_005254013.2 | XP_005254070.1 | 3 - 4 | 917 | 81 | |
XM_006719717.2 | XP_006719780.1 | 2 - 3 | 860 | 81 | |
XM_017020282.1 | XP_016875771.1 | 3 - 4 | 917 | 81 | |
XM_017020283.1 | XP_016875772.1 | 2 - 3 | 860 | 81 | |
GenBank mRNA | AB027196.1 | - | 2 - 3 | 804 | 81 |
AK094011.1 | - | 3 - 4 | 741 | 81 | |
AK097807.1 | - | 3 - 4 | 766 | 81 | |
AK126274.1 | - | 3 - 4 | 645 | 81 | |
AK301608.1 | - | 3 - 4 | 714 | 81 | |
AK315558.1 | - | 2 - 3 | 822 | 81 | |
AL389976.1 | - | 1 - 2 | 97 | 81 | |
BC016622.1 | - | 2 - 3 | 804 | 81 | |
BC031596.1 | - | 2 - 3 | 804 | 81 | |
BC101709.1 | - | 2 - 3 | 430 | 81 | |
BC101715.1 | - | 2 - 3 | 430 | 81 | |
BC143493.1 | - | 2 - 3 | 429 | 81 | |
BT006974.1 | - | 2 - 3 | 357 | 81 | |
D87451.2 | - | 2 - 3 | 357 | 81 |