Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013251.3 | NP_037383.1 | 4 - 5 | 418 | 75 |
NR_033654.1 | 4 - 5 | 418 | 75 | ||
NR_135164.1 | 4 - 5 | 418 | 75 | ||
NR_135165.1 | 4 - 5 | 418 | 75 | ||
GenBank mRNA | AF186112.1 | - | 4 - 5 | 361 | 75 |
AF216586.1 | - | 4 - 5 | 348 | 75 | |
AF537113.1 | - | 4 - 5 | 378 | 75 | |
AF537114.1 | - | 4 - 5 | 378 | 75 | |
AF537115.1 | - | 4 - 5 | 378 | 75 | |
AF537116.1 | - | 4 - 5 | 378 | 75 | |
AF537117.1 | - | 4 - 5 | 378 | 75 | |
AF537118.1 | - | 4 - 5 | 378 | 75 | |
AX147032.1 | - | 4 - 5 | 264 | 75 | |
AY358679.1 | - | 4 - 5 | 396 | 75 | |
BC032145.2 | - | 4 - 5 | 399 | 75 | |
BP212420.1 | - | 4 - 5 | 418 | 75 | |
CR457193.1 | - | 3 - 4 | 239 | 75 |