Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178054.1 | NP_001171525.1 | 5 - 6 | 494 | 93 |
NM_013251.3 | NP_037383.1 | 6 - 7 | 548 | 93 | |
NR_033654.1 | 7 - 8 | 665 | 93 | ||
GenBank mRNA | AF186112.1 | - | 6 - 7 | 491 | 93 |
AF216586.1 | - | 6 - 7 | 478 | 93 | |
AF537115.1 | - | 7 - 8 | 625 | 93 | |
AF537118.1 | - | 6 - 7 | 508 | 93 | |
AF537121.1 | - | 5 - 6 | 454 | 93 | |
AX147032.1 | - | 6 - 7 | 394 | 93 | |
AY358679.1 | - | 7 - 8 | 643 | 93 | |
BC032145.2 | - | 6 - 7 | 529 | 93 |