Assay Design



Gene Transcripts

Gene Symbol
NADSYN1
Entrez Gene ID
55191
Gene Name
NAD synthetase 1
Gene Aliases
-
Chromosome Location
Chr.11: 71453171 - 71501538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018161.4 NP_060631.2 8 - 9 795 103
GenBank mRNA AB091316.1 - 8 - 9 669 103
AK001493.1 - 8 - 9 736 103
AK022436.1 - 8 - 9 765 103
AK225700.1 - 8 - 9 749 103
BC003638.2 - 8 - 9 758 103
BC003666.2 - 8 - 9 758 103
CR457286.1 - 8 - 9 669 103

View Full Product Details