Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080448.2 | NP_001073917.2 | 16 - 17 | 3176 | 134 |
XM_006713592.3 | XP_006713655.1 | 17 - 18 | 3224 | 134 | |
XM_017006210.1 | XP_016861699.1 | 16 - 17 | 3098 | 134 | |
XM_017006211.1 | XP_016861700.1 | 18 - 19 | 3249 | 134 | |
XM_017006212.1 | XP_016861701.1 | 17 - 18 | 2918 | 134 | |
XM_017006213.1 | XP_016861702.1 | 17 - 18 | 3276 | 134 | |
XM_017006214.1 | XP_016861703.1 | 16 - 17 | 2544 | 134 | |
XM_017006215.1 | XP_016861704.1 | 12 - 13 | 1573 | 134 | |
XM_017006216.1 | XP_016861705.1 | 11 - 12 | 1523 | 134 | |
XM_017006217.1 | XP_016861706.1 | 13 - 14 | 1620 | 134 | |
XM_017006218.1 | XP_016861707.1 | 11 - 12 | 1423 | 134 | |
XM_017006220.1 | XP_016861709.1 | 13 - 14 | 1588 | 134 | |
XM_017006221.1 | XP_016861710.1 | 11 - 12 | 1534 | 134 | |
XM_017006222.1 | XP_016861711.1 | 12 - 13 | 1582 | 134 | |
GenBank mRNA | DR731365.1 | - | 1 - 2 | 117 | 134 |