Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080448.2 | NP_001073917.2 | 4 - 5 | 1317 | 133 |
XM_006713592.3 | XP_006713655.1 | 4 - 5 | 1317 | 133 | |
XM_017006210.1 | XP_016861699.1 | 4 - 5 | 1317 | 133 | |
XM_017006211.1 | XP_016861700.1 | 5 - 6 | 1342 | 133 | |
XM_017006212.1 | XP_016861701.1 | 4 - 5 | 1011 | 133 | |
XM_017006213.1 | XP_016861702.1 | 4 - 5 | 1369 | 133 | |
XM_017006214.1 | XP_016861703.1 | 3 - 4 | 637 | 133 | |
XM_017006219.1 | XP_016861708.1 | 4 - 5 | 1317 | 133 | |
GenBank mRNA | AK092565.1 | - | 4 - 5 | 1312 | 133 |