Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286682.1 | NP_001273611.1 | 8 - 9 | 935 | 140 |
NM_001286683.1 | NP_001273612.1 | 8 - 9 | 1008 | 140 | |
NM_016228.3 | NP_057312.1 | 7 - 8 | 1141 | 140 | |
NM_182662.1 | NP_872603.1 | 8 - 9 | 923 | 140 | |
XM_006714231.2 | XP_006714294.1 | 9 - 10 | 1065 | 140 | |
XM_011532020.2 | XP_011530322.1 | 5 - 6 | 698 | 140 | |
GenBank mRNA | AF481738.1 | - | 7 - 8 | 1142 | 140 |
AK055952.1 | - | 8 - 9 | 923 | 140 | |
AK314729.1 | - | 8 - 9 | 1010 | 140 | |
BC031068.1 | - | 8 - 9 | 928 | 140 |