Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308028.1 | NP_001294957.1 | 2 - 3 | 215 | 73 |
NM_005246.3 | NP_005237.2 | 2 - 3 | 327 | 73 | |
XM_011543266.1 | XP_011541568.1 | 2 - 3 | 180 | 73 | |
XM_011543267.1 | XP_011541569.1 | 2 - 3 | 2064 | 73 | |
XM_011543271.2 | XP_011541573.1 | 2 - 3 | 180 | 73 | |
XM_011543272.1 | XP_011541574.1 | 2 - 3 | 180 | 73 | |
XM_011543278.1 | XP_011541580.1 | 2 - 3 | 180 | 73 | |
XM_017009234.1 | XP_016864723.1 | 2 - 3 | 180 | 73 | |
XM_017009235.1 | XP_016864724.1 | 2 - 3 | 180 | 73 | |
GenBank mRNA | AK299855.1 | - | 2 - 3 | 180 | 73 |
AL707447.1 | - | 2 - 3 | 215 | 73 | |
J03358.1 | - | 2 - 3 | 327 | 73 |