Assay Design



Gene Transcripts

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
Chromosome Location
Chr.3: 96814581 - 97761532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006210.1 XP_016861699.1 12 - 13 2478 114
GenBank mRNA AK310249.1 - 7 - 8 761 114

View Full Product Details