Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015559.2 | NP_056374.2 | 5 - 6 | 4469 | 111 |
GenBank mRNA | AB007897.2 | - | 5 - 6 | 4469 | 111 |
AB022660.1 | - | 4 - 5 | 4016 | 111 | |
BC146776.1 | - | 5 - 6 | 4469 | 111 | |
BQ641251.1 | - | 2 - 3 | 565 | 111 |