Assay Design
Gene Transcripts
- Gene Symbol
- COL4A2
- Entrez Gene ID
-
1284
- Gene Name
- collagen type IV alpha 2
- Gene Aliases
-
ICH, POREN2
- Chromosome Location
-
Chr.13: 110307284 - 110513027 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CB045156.1 |
- |
3 - 4 |
159 |
82 |
X05562.1 |
- |
12 - 13 |
1020 |
82 |
View Full Product Details