Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AF258350.1 | - | 2 - 3 | 432 | 65 |
AF400430.1 | - | 2 - 3 | 432 | 65 | |
AK025912.1 | - | 15 - 16 | 1992 | 65 | |
AK294920.1 | - | 19 - 20 | 2319 | 65 | |
AY450357.1 | - | 2 - 3 | 432 | 65 | |
AY455978.1 | - | 2 - 3 | 432 | 65 | |
BC080644.1 | - | 5 - 6 | 839 | 65 | |
BX648393.1 | - | 16 - 17 | 2058 | 65 | |
J02760.1 | - | 3 - 4 | 537 | 65 | |
X05610.1 | - | 7 - 8 | 1128 | 65 |