Assay Design



Gene Transcripts

Gene Symbol
TMEM38A
Entrez Gene ID
79041
Gene Name
transmembrane protein 38A
Gene Aliases
TRIC-A, TRICA
Chromosome Location
Chr.19: 16661127 - 16689020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024074.2 NP_076979.1 5 - 6 762 94
GenBank mRNA AK025981.1 - 5 - 6 762 94
AK292764.1 - 5 - 6 747 94
BC001195.1 - 5 - 6 678 94

View Full Product Details