Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204747.1 | NP_001191676.1 | 3 - 4 | 354 | 143 |
NM_002913.4 | NP_002904.3 | 3 - 4 | 354 | 143 | |
XM_011513730.1 | XP_011512032.1 | 3 - 4 | 354 | 143 | |
XM_011513731.1 | XP_011512033.1 | 3 - 4 | 354 | 143 | |
XM_017008528.1 | XP_016864017.1 | 3 - 4 | 354 | 143 | |
GenBank mRNA | AF040250.1 | - | 3 - 4 | 259 | 143 |
AK291612.1 | - | 3 - 4 | 335 | 143 | |
BC010387.1 | - | 4 - 5 | 460 | 143 | |
BC035297.1 | - | 3 - 4 | 305 | 143 | |
BC051751.1 | - | 3 - 4 | 324 | 143 | |
BC051786.2 | - | 3 - 4 | 335 | 143 | |
DA336360.1 | - | 3 - 4 | 354 | 143 | |
L23320.1 | - | 3 - 4 | 420 | 143 |