Assay Design
Gene Transcripts
- Gene Symbol
- DNAJC22
- Entrez Gene ID
-
79962
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C22
- Gene Aliases
-
wus
- Chromosome Location
-
Chr.12: 49346917 - 49353684 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024902.3 |
NP_079178.2 |
1 - 2 |
637 |
67 |
GenBank mRNA |
BC033236.1 |
- |
1 - 2 |
296 |
67 |
View Full Product Details