Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031469.3 | NP_113657.1 | 1 - 2 | 406 | 76 |
XM_017011344.1 | XP_016866833.1 | 3 - 4 | 5446 | 76 | |
GenBank mRNA | AF340151.1 | - | 1 - 2 | 228 | 76 |
AJ297972.1 | - | 1 - 2 | 49 | 76 | |
AK074506.1 | - | 1 - 2 | 208 | 76 | |
AK311757.1 | - | 1 - 2 | 214 | 76 | |
BC040489.1 | - | 1 - 2 | 214 | 76 | |
BC052987.1 | - | 1 - 2 | 206 | 76 | |
BC060799.1 | - | 1 - 2 | 182 | 76 | |
BC109043.2 | - | 1 - 2 | 216 | 76 | |
BC109044.2 | - | 1 - 2 | 216 | 76 | |
DC417547.1 | - | 1 - 2 | 406 | 76 |