Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031469.3 | NP_113657.1 | 2 - 3 | 585 | 81 |
XM_011536165.2 | XP_011534467.1 | 2 - 3 | 559 | 81 | |
XM_017011344.1 | XP_016866833.1 | 4 - 5 | 5625 | 81 | |
GenBank mRNA | AF340151.1 | - | 2 - 3 | 407 | 81 |
AJ297972.1 | - | 2 - 3 | 228 | 81 | |
AK311757.1 | - | 2 - 3 | 393 | 81 | |
BC040489.1 | - | 2 - 3 | 393 | 81 | |
BC052987.1 | - | 2 - 3 | 385 | 81 | |
BC060799.1 | - | 2 - 3 | 361 | 81 | |
BC070059.1 | - | 2 - 3 | 361 | 81 | |
BC109043.2 | - | 2 - 3 | 395 | 81 | |
BC109044.2 | - | 2 - 3 | 395 | 81 |