Assay Design



Gene Transcripts

Gene Symbol
HTRA3
Entrez Gene ID
94031
Gene Name
HtrA serine peptidase 3
Gene Aliases
Prsp, Tasp
Chromosome Location
Chr.4: 8269757 - 8307111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297559.1 NP_001284488.1 3 - 4 914 62
NM_053044.4 NP_444272.1 3 - 4 914 62
XM_011513596.2 XP_011511898.1 3 - 4 922 62
GenBank mRNA AL547377.3 - 3 - 4 917 62
AY040094.2 - 3 - 4 931 62
AY280665.1 - 3 - 4 902 62
AY280666.1 - 3 - 4 817 62
BC034390.1 - 3 - 4 914 62
BC035717.1 - 3 - 4 914 62

View Full Product Details