Assay Design



Gene Transcripts

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
Chromosome Location
Chr.7: 7338543 - 7542062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011515360.2 XP_011513662.1 35 - 36 4782 125

View Full Product Details