Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282349.1 | NP_001269278.1 | 4 - 5 | 563 | 111 |
NM_001282350.1 | NP_001269279.1 | 3 - 4 | 530 | 111 | |
NM_130771.4 | NP_570127.3 | 5 - 6 | 575 | 111 | |
NM_133168.4 | NP_573398.2 | 3 - 4 | 530 | 111 | |
NM_133169.4 | NP_573399.2 | 4 - 5 | 563 | 111 | |
NM_206818.2 | NP_996554.2 | 5 - 6 | 575 | 111 | |
GenBank mRNA | AF251702.1 | - | 3 - 4 | 417 | 111 |
AF391162.1 | - | 3 - 4 | 378 | 111 | |
AF391163.1 | - | 4 - 5 | 383 | 111 | |
AF391164.1 | - | 2 - 3 | 376 | 111 | |
AF474152.1 | - | 3 - 4 | 409 | 111 | |
AF474153.1 | - | 2 - 3 | 338 | 111 | |
AK091111.1 | - | 3 - 4 | 380 | 93 | |
AK130199.1 | - | 4 - 5 | 407 | 111 | |
AK311307.1 | - | 3 - 4 | 503 | 111 | |
BC035023.2 | - | 4 - 5 | 564 | 111 | |
BG722509.1 | - | 4 - 5 | 569 | 111 |