Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020533.2 | NP_065394.1 | 9 - 10 | 1272 | 63 |
GenBank mRNA | AF249319.1 | - | 9 - 10 | 1215 | 63 |
AF287269.1 | - | 9 - 10 | 1256 | 63 | |
AJ293659.1 | - | 8 - 9 | 1446 | 63 | |
AJ293970.1 | - | 9 - 10 | 1211 | 63 | |
AK026102.1 | - | 9 - 10 | 1226 | 63 | |
AK222673.1 | - | 9 - 10 | 1251 | 63 | |
AK294330.1 | - | 9 - 10 | 1237 | 63 | |
BC005149.2 | - | 9 - 10 | 1254 | 63 |