Assay Design



Gene Transcripts

Gene Symbol
CTSW
Entrez Gene ID
1521
Gene Name
cathepsin W
Gene Aliases
LYPN
Chromosome Location
Chr.11: 65879813 - 65883741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335.3 NP_001326.2 1 - 2 123 86
GenBank mRNA AF013611.1 - 1 - 2 81 86
BC035637.1 - 1 - 2 86 86
BC048255.1 - 1 - 2 123 86
BI833187.1 - 1 - 2 123 86

View Full Product Details